Learn More
Chemglass Life Sciences Connector, Lattice
SDP
Catalog No. 50871984
Encompass_Preferred
Connector, Lattice. Angled adjustment screws allow easy set-up and prevent slippage or misalignment. Two U-shaped slots permit rods or clamps of 1/2” (13mm) maximum diameter to be joined in a 90° corner joint or cross-connection with components in close proximity to minimize wasted space. Each rod or clamp adjusts independently for easy installation.
Catalog No. 50-871-984 Supplier Chemglass Life Sciences Supplier No. CG926001
Connector, Lattice. Angled adjustment screws allow easy set-up and prevent slippage or misalignment. Two U-shaped slots permit rods or clamps of 1/2” (13mm) maximum diameter to be joined in a 90° corner joint or cross-connection with components in close proximity to minimize wasted space. Each rod or clamp adjusts independently for easy installation.