Learn More
Metrohm USA CONNECTOR METROHM
SDP
Supplier:  Metrohm USA 61808090
Encompass
Outer thread M8, inner thread M6, e.g. for mounting 6.2418.XXX Air tubes in 6.1429.XXX Reaction vessels of stability measuring instruments.
Catalog No. NC9904071