Learn More
Grainger Connector RJ45/180 Degree 24AWG
Catalog No. 502749279
Encompass_Preferred
The Siemens 14G833 is an RJ45 connector designed for industrial Ethernet applications. This unshielded connector has dimensions of 45x45x27mm and features a robust metal housing with 8 gold plated contacts. It is IP20 rated for protection against solid objects greater than 12mm. The operating temperature range is -40 to 85°C. This connector supports 10/100/1000 Mbps data rates and meets IEC 60603-7 standards. The max current rating is 1.5A at 60V DC. It has 360 degree shielding and plug retention force of 4.4 lb. This RJ45 connector comes in a pack of 1 and has a 2 year warranty.
Catalog No. 50-274-9279 Supplier Grainger Supplier No. 14G833
The Siemens 14G833 is an RJ45 connector designed for industrial Ethernet applications. This unshielded connector has dimensions of 45x45x27mm and features a robust metal housing with 8 gold plated contacts. It is IP20 rated for protection against solid objects greater than 12mm. The operating temperature range is -40 to 85°C. This connector supports 10/100/1000 Mbps data rates and meets IEC 60603-7 standards. The max current rating is 1.5A at 60V DC. It has 360 degree shielding and plug retention force of 4.4 lb. This RJ45 connector comes in a pack of 1 and has a 2 year warranty.