Learn More
Chemglass Life Sciences S-Connector
SDP
Catalog No. 50871986
Encompass_Preferred
S-Connector. Similar to the CG-9260-01 lattice connector, the “S” connector features ergonomic contouring for easy adjustment with angled set screws for maximum grip against rods or clamps.
Catalog No. 50-871-986 Supplier Chemglass Life Sciences Supplier No. CG926401
S-Connector. Similar to the CG-9260-01 lattice connector, the “S” connector features ergonomic contouring for easy adjustment with angled set screws for maximum grip against rods or clamps.