Learn More
Chemglass Life Sciences Complete Kit: Threaded Vial Adapters Only: 13-425, 15-425, 18-425, 20-400, 22-400, 24-400
SDP
Catalog No. CG131830
Encompass_Preferred
Rotary adapter kit. Adapters permit the connection of standard laboratory dram sample vials directly to the vapor tube or a bump trap of a rotary evaporator. Adapters have a top outer joint of the listed size and a 20-400 G.P.I. thread at the bottom. A PTFE-lined, polypropylene, double-threaded connector attaches the vial to the adapter. Threaded Vial Adapters Only: 13-425, 15-425, 18-425, 20-400, 22-400, 24-400.
Catalog No. CG131830 Supplier Chemglass Life Sciences Supplier No. CG131830
Rotary adapter kit. Adapters permit the connection of standard laboratory dram sample vials directly to the vapor tube or a bump trap of a rotary evaporator. Adapters have a top outer joint of the listed size and a 20-400 G.P.I. thread at the bottom. A PTFE-lined, polypropylene, double-threaded connector attaches the vial to the adapter. Threaded Vial Adapters Only: 13-425, 15-425, 18-425, 20-400, 22-400, 24-400.