Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, 316 Stainless Steel, Straight, Hose Barb Union, 1/2"" ID
Catalog No. 50629280
Encompass
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Catalog No. 50-629-280 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3062006
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.