Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, PVDF, Straight, Hose Barb Union, 3/8" ID; 10/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3070307/DUP
Encompass
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Catalog No. 50-629-596