Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, 316 Stainless Steel, Straight, Hose Barb Union, 5/8"" ID
Catalog No. 50629109
Encompass
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Catalog No. 50-629-109 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3062007
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.