Learn More
Med Vet International Dynarex Tracheostomy Pediatric Flex Connector, 15 mm Od X 15 mm Id, 50/Case
Catalog No. 502829690
Encompass_Preferred
Dynarex Resp-O2 Flex Tube/Flex Connectors add length to vertical circuits and HMEs to help improve system flexibility and range of motion. Provides additional flexibility to ventilator circuits Lightweight, pliable design Not made with natural rubber latex or DEHP
Catalog No. 50-282-9690 Supplier Med Vet International Supplier No. 30602
Dynarex Resp-O2 Flex Tube/Flex Connectors add length to vertical circuits and HMEs to help improve system flexibility and range of motion. Provides additional flexibility to ventilator circuits Lightweight, pliable design Not made with natural rubber latex or DEHP