Learn More
Glass Expansion Connector Tube with 1/4-28 fitting 0.5mm x 700mm (PKT 3)
SDP
Catalog No. 502703030
Encompass_Preferred
Connector Tube with 1/4-28 fitting 0.5mm x 700mm (PKT 3). A pack of 3 replacement sample connector tubes for the Trident CT Internal Standard Kit (P/Ns NC2046960, 60-703-1179).Replacement ferrules 70-803-0749 and ratchet connectors 70-803-1350.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3030 Supplier Glass Expansion Supplier No. 708031871
Connector Tube with 1/4-28 fitting 0.5mm x 700mm (PKT 3). A pack of 3 replacement sample connector tubes for the Trident CT Internal Standard Kit (P/Ns NC2046960, 60-703-1179).Replacement ferrules 70-803-0749 and ratchet connectors 70-803-1350.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001