Learn More
SOMNI Scientific™ Connector Vaporizer Endcap Set
SDP
Catalog No. 04777230 Shop All Somni Scientific Products
Change view
Click to view available options
Size:
23 mm
Catalog No. Size
04-777-230 23 mm
1 options
Catalog No. 04-777-230 Supplier SOMNI Scientific™ Supplier No. VP6211
Add to Cart
Add to Cart
The Connector Vaporizer Endcap Set is a stainless steel connector that provides a snug connection from the oxygen source to the inlet of the vaporizer and from the outlet of the vaporizer to the anesthesia system.
  • 1/4 in. hosebarb accepts 3/16 in.ID silicone tubing
Specifications
For Use With (Application) Animal Inhalant Anesthesia
Disposable Reusable
For Use With (Equipment) 3/16 in. ID Silicone Tubing, Oxygen Source, Vaporizer, Anesthesia System
Size 23 mm
Sterility Non-sterile