Learn More
MilliporeSigma™ Connectors for Milli-Q™ IQ 7000 Ultrapure Water System
Shop All MilliporeSigma Products

Description
Order Info
Must be ordered with the Milli-Q™ IQ 7000 Ultrapure Water System. Please contact your Fisher Rep for more details.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| For Use With (Equipment) | Milli-Q IQ 7000 Ultrapure Water System |
| Type | Connector 5m Q-POD to Q-POD |
| Purity Meter Included | No |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.