Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Connectors for Milli-Q™ IQ 7000 Ultrapure Water System
GSA_VA

Catalog No. ZFC0NN5SQ Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Connector 2m Q-POD to Q-POD
Connector 2m System to Q-POD
Connector 5m Q-POD to Q-POD
Connector 5m System to Q-POD
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
ZFC0NN5SQ Connector 5m System to Q-POD
ZFC0NN2QQ Connector 2m Q-POD to Q-POD
ZFC0NN2SQ Connector 2m System to Q-POD
ZFC0NN5QQ Connector 5m Q-POD to Q-POD
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ZFC0NN5SQ Supplier MilliporeSigma™ Supplier No. ZFC0NN5SQ
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The most integrated pure and ultrapure water system that is intelligent, intuitive and green.

Order Info

Must be ordered with the Milli-Q™ IQ 7000 Ultrapure Water System. Please contact your Fisher Rep for more details.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Milli-Q IQ 7000 Ultrapure Water System
Type Connector 5m System to Q-POD
Purity Meter Included No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.