Learn More
AVANTOR MASTERFLEX (VWR LLC) Barbed fittings, Straight Connector, Natural PP, 1/2"" ID,11/32"", 2-21/32"", 11/16""
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX4061003/DEL
Encompass
Barbed fittings, Straight Connector, Natural PP, 1/2" ID, 10/Pk, Quick and easy assembly without toolsIdeal for low-pressure applicationsChemical resistantAvailable in a variety of sizes, materials, and configurations
Catalog No. NC0894982