Learn More
Bioss Connexin-43 C-terminal region
SDP
Catalog No. 5000065728
Encompass_Preferred
Connexin-43 (C-terminal region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Chicken Clonality Monoclonal Applications WB M496Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-65728 Supplier Bioss Supplier No. BSM70425M
Connexin-43 (C-terminal region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Chicken Clonality Monoclonal Applications WB M496Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol