Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Conoidin A 10mM 1mL | 18080-67-6 | 347.99 g/mol | C10H8Br2N2O2 | 1 ML
SDP

Catalog No. 5000229751
Encompass_Preferred

Conoidin A is a cell-permeable, covalent inhibitor of peroxiredoxins provided as a ready-to-use 10 mM solution (1 mL) in DMSO for research applications. It is used in biochemical and cell-based studies of oxidative stress and parasite enzyme inhibition and is supplied with defined purity and storage recommendations.

  • 10 mM solution, 1 mL, in DMSO.
  • Purity 99.32% as listed by supplier.
  • CAS 18080-67-6.
  • Molecular formula C10H8Br2N2O2; molecular weight 347.99 g/mol.
  • Intended for research use only; not for human or clinical use.
  • Storage: in solvent -80 °C (up to 6 months) or -20 °C (up to 1 month).

Catalog No. 50-002-29751 Supplier Medchemexpress LLC Supplier No. HY11609010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Conoidin A is a cell-permeable, covalent inhibitor of peroxiredoxins provided as a ready-to-use 10 mM solution (1 mL) in DMSO for research applications. It is used in biochemical and cell-based studies of oxidative stress and parasite enzyme inhibition and is supplied with defined purity and storage recommendations.

  • 10 mM solution, 1 mL, in DMSO.
  • Purity 99.32% as listed by supplier.
  • CAS 18080-67-6.
  • Molecular formula C10H8Br2N2O2; molecular weight 347.99 g/mol.
  • Intended for research use only; not for human or clinical use.
  • Storage: in solvent -80 °C (up to 6 months) or -20 °C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.