Learn More
Cayman Chemical Conoidin A, 18080-67-6, 25 mg
SDP
Supplier:  Cayman Chemical 1560525
Encompass_Preferred
An inhibitor of parasitic and mammalian peroxiredoxins (T. gondii peroxiredoxin II (IC50 = 23 µM), A. ceylanicum peroxiredoxin I, and mammalian peroxiredoxin I and II (5 µM))
Catalog No. NC0676161