Learn More
SCP Science Standard S-21, 10 ppm, 200 g
Catalog No. NC0118078
Encompass
Oil based metal calibration standards for use with ICP, AA, rotrode, XRF, flame emission and other instruments. S-21: Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn.
Catalog No. NC0118078 Supplier SCP Science Supplier No. 150021018
Oil based metal calibration standards for use with ICP, AA, rotrode, XRF, flame emission and other instruments. S-21: Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn.