Learn More
LAMDA BIOTECH INC Conquest Genotyping PCR Kit
Catalog No. NC3724385
Encompass_Preferred
A comprehensive system combining genomic DNA extraction reagents with four distinct 2X PCR Master Mixes for genotyping applications. This kit enables rapid DNA extraction from diverse biological sources followed by direct PCR amplification.
Catalog No. NC3724385 Supplier LAMDA BIOTECH INC Supplier No. D911
A comprehensive system combining genomic DNA extraction reagents with four distinct 2X PCR Master Mixes for genotyping applications. This kit enables rapid DNA extraction from diverse biological sources followed by direct PCR amplification.