Learn More
Hach Company Const. Effluent Monit. Kit
Supplier:  Hach Company 2527100
Encompass
Construction Effluent Monitoring KitIncludes 2100Q Portable Turbidimeter (2100Q01)-Pocket Pro pH Tester (9532000)-SINGLET pH buffer combination pack (pH 4 01 7 00) (2769920)-2 x 120 mL sample containers (2552200)-250 mL wash bottle (62031) for rinse water-and a hard-sided carrying case (4660500) Kit for monitoring pH and turbidity of construction site stormwater effluent Ideal for compliance monitoring in states such as California and Washington with numeric limits included in the Construction General Permit
Catalog No. 50-003-04271