Learn More
World Precision Instrument Constant Current Stimulus Isolator-SYS-A365D
SDP
Catalog No. 502538504
Encompass
Activated by conventional logic-level commands, Model A365 can be gated by any pulse generator, stimulator or computer output with automated bipolar pulsing for zero net charge on biological preparations.
To learn more about our warranty options, click here.
Catalog No. 50-253-8504 Supplier World Precision Instrument Supplier No. SYSA365D
Activated by conventional logic-level commands, Model A365 can be gated by any pulse generator, stimulator or computer output with automated bipolar pulsing for zero net charge on biological preparations.
To learn more about our warranty options, click here.