Learn More
MSC Value Collection Heat Crimper, 1/2 x 1/4 in, 1 ea
Catalog No. 5000795184
Encompass
This handheld heat crimper is designed for sealing applications. It operates at 60 W and 110 V, making it a reliable tool for various tasks.
  • Handheld heat crimper type
  • Maximum seal size of 1/2 x 1/4 in
  • Operates at 60 W
  • Operates at 110 V
  • Manufactured in the United States
Catalog No. 50-007-95184 Supplier MSC Supplier No. 76219013
This handheld heat crimper is designed for sealing applications. It operates at 60 W and 110 V, making it a reliable tool for various tasks.
  • Handheld heat crimper type
  • Maximum seal size of 1/2 x 1/4 in
  • Operates at 60 W
  • Operates at 110 V
  • Manufactured in the United States