Learn More
Sigma Aldrich Constantan - resistance alloy, Cu55Ni45
Catalog No. 503097130 Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
50-309-7130 1 Ea.
1 options
Catalog No. 50-309-7130 Supplier Sigma Aldrich Supplier No. GF529351721EA
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Constantan - resistance alloy, Cu55Ni45
Specifications
Quantity 1 Ea.
Synonym CU045815