Learn More
Sony Biotechnology Consumable, single-use sterile QC beads for use with the CGX10 Cell Isolation System -
Catalog No. NC3885091
Encompass_Preferred
Consumable, single-use sterile QC beads for use with the CGX10 Cell Isolation System - Contents: 3 bags, each containing 2 vials
Vial 1
• Capacity: 1.5 mL
• 10 μm beads: 2.5 × 106/mL
Vial 2
• Capacity: 1 mL
• 3 μm beads: 1.0 × 107/mL
Catalog No. NC3885091 Supplier Sony Biotechnology Supplier No. LECGXQC1
Consumable, single-use sterile QC beads for use with the CGX10 Cell Isolation System - Contents: 3 bags, each containing 2 vials
Vial 1
• Capacity: 1.5 mL
• 10 μm beads: 2.5 × 106/mL
Vial 2
• Capacity: 1 mL
• 3 μm beads: 1.0 × 107/mL