Learn More
Thermo Scientific™ Consumables for KingFisher™ mL Systems
Catalog No. 22387026 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 45 pieces
800 pieces
For 240 samples
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-387-026 For 240 samples
22-387-024 800 pieces
22-387-025 20 x 45 pieces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-387-026 Supplier Thermo Scientific™ Supplier No. 97002141
Add to Cart
Add to Cart
Optimize system performance with Consumables for Thermo Scientific™ KingFisher™ mL Systems.
Optimize system performance with Consumables for Thermo Scientific™ KingFisher™ mL Systems.
These tube strips are specifically designed for use with the Thermo Scientific™ KingFisher™ mL instrument, ensuring compatibility with the system.
The KingFisher mL instrument is designed for a maximum of 15 tube strips, which are compatible with the specially desinged tip comb.
  • One sample processing uses one tube strip containing five tubes.
  • One tip comb with five tips is used for processing five samples at a time.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) KingFisher mL Combi 240
Quantity For 240 samples
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.