Learn More
Perkin Elmer US LLC Consumables Kit for NexION 2000 Productivity Configuration
Catalog No. 502103960
Encompass_Preferred
This kit includes: Demountable Quartz Torch - Black Mark, Qty.2 (N8152379) 2.0 mm Demountable Quartz Injector - Black Mark, Qty.2 (N8152376) Torch Injector Nut, Qty.2 (N8152451) Glass Cyclonic High Sensitivity Spray Chamber with Matrix Gas Port, Qty.1 (N8152389) Threaded Spray Chamber Drain Line, Qty.2 (N8152456) Nickel Skimmer Cone, Qty.2 (W1026356) Nickel Sampler Cone, Qty.2 (W1033612) Hyper Skimmer Cone, Qty.1 (W1033995) Sampler Cone Gasket, Qty.3 (W1040148) Hyper Skimmer O-Ring, Qty.2 (09902123) Gray/Gray Pump Tubing MP2, Qty.2 (N8145173) Flared PVC 0.51 mm Orange/Yellow Pump Tubing MP2, Qty.4 (N8145150) Drain Tubing (feet), Qty.6 (02506495) Sample Uptake Tubing, Qty.12 (WE024375) Sample Introduction Line for PFA-ST Nebulizer, Qty.1 (N8152455) Female Barb CTFE Fitting, Qty.4 (N8152388) Male Barb CTFE Fitting, Qty.4 (N8152453) Instrument Filter (Left), Qty.2 (N8152167) Instrument Filter (Right), Qty.2 (N8152136)
Catalog No. 50-210-3960 Supplier Perkin Elmer US LLC Supplier No. N8150056
Add to Cart
Add to Cart
Request a Quote
This kit includes: Demountable Quartz Torch - Black Mark, Qty.2 (N8152379) 2.0 mm Demountable Quartz Injector - Black Mark, Qty.2 (N8152376) Torch Injector Nut, Qty.2 (N8152451) Glass Cyclonic High Sensitivity Spray Chamber with Matrix Gas Port, Qty.1 (N8152389) Threaded Spray Chamber Drain Line, Qty.2 (N8152456) Nickel Skimmer Cone, Qty.2 (W1026356) Nickel Sampler Cone, Qty.2 (W1033612) Hyper Skimmer Cone, Qty.1 (W1033995) Sampler Cone Gasket, Qty.3 (W1040148) Hyper Skimmer O-Ring, Qty.2 (09902123) Gray/Gray Pump Tubing MP2, Qty.2 (N8145173) Flared PVC 0.51 mm Orange/Yellow Pump Tubing MP2, Qty.4 (N8145150) Drain Tubing (feet), Qty.6 (02506495) Sample Uptake Tubing, Qty.12 (WE024375) Sample Introduction Line for PFA-ST Nebulizer, Qty.1 (N8152455) Female Barb CTFE Fitting, Qty.4 (N8152388) Male Barb CTFE Fitting, Qty.4 (N8152453) Instrument Filter (Left), Qty.2 (N8152167) Instrument Filter (Right), Qty.2 (N8152136)