Learn More
Perkin Elmer US LLC Consumables Kit for NexION 2000 Base Configuration
Catalog No. 502103959
Encompass_Preferred
This kit includes: One-Piece Quartz Torch, 2.0 mm I.D. Injector - Blue/Purple Mark, Qty.2 (N8152472) Torch Injector Nut, Qty.2 (N8152451) Glass Cyclonic High Sensitivity Spray Chamber, Qty.1 (N8152375) Threaded Spray Chamber Drain Line, Qty.2 (N8152456) Nickel Skimmer Cone, Qty.2 (W1026356) Nickel Sampler Cone, Qty.2 (W1033612) Hyper Skimmer Cone, Qty.1 (W1033995) Sampler Cone Gasket, Qty.3 (W1040148) Hyper Skimmer O-Ring, Qty.2 (09902123) Gray/Gray Pump Tubing, Qty.2 (N8152415) Flared PVC 0.51 mm I.D. Orange/Yellow Pump Tubing, Qty.4 (N8152405) Drain Tubing (feet), Qty. 6 (02506495) Sample Uptake Tubing, Qty. 12 (WE024375) Sample Introduction Line for Concentric Nebulizer, Qty.1 (N8145503) Female Barb CTFE Fitting, Qty.4 (N8152388) Male Barb CTFE Fitting, Qty.4 (N8152453) Instrument Filter (Left), ty.2 (N8152167) Instrument Filter (Right) qty.2 (N8152136)
Catalog No. 50-210-3959 Supplier Perkin Elmer US LLC Supplier No. N8150054
Add to Cart
Add to Cart
Request a Quote
This kit includes: One-Piece Quartz Torch, 2.0 mm I.D. Injector - Blue/Purple Mark, Qty.2 (N8152472) Torch Injector Nut, Qty.2 (N8152451) Glass Cyclonic High Sensitivity Spray Chamber, Qty.1 (N8152375) Threaded Spray Chamber Drain Line, Qty.2 (N8152456) Nickel Skimmer Cone, Qty.2 (W1026356) Nickel Sampler Cone, Qty.2 (W1033612) Hyper Skimmer Cone, Qty.1 (W1033995) Sampler Cone Gasket, Qty.3 (W1040148) Hyper Skimmer O-Ring, Qty.2 (09902123) Gray/Gray Pump Tubing, Qty.2 (N8152415) Flared PVC 0.51 mm I.D. Orange/Yellow Pump Tubing, Qty.4 (N8152405) Drain Tubing (feet), Qty. 6 (02506495) Sample Uptake Tubing, Qty. 12 (WE024375) Sample Introduction Line for Concentric Nebulizer, Qty.1 (N8145503) Female Barb CTFE Fitting, Qty.4 (N8152388) Male Barb CTFE Fitting, Qty.4 (N8152453) Instrument Filter (Left), ty.2 (N8152167) Instrument Filter (Right) qty.2 (N8152136)