Learn More
Perkin Elmer US LLC Consumables Kit for NexION 300/350 D/Q/X
Catalog No. 502104760
Encompass_Preferred
This kit includes: Demountable Quartz Torch, Qty. 2 (N8122006) 2 mm I.D. Quartz Injector with Ball Joint, Qty. 2 (WE023948) Glass Cyclonic Spray Chamber, Qty. 1, (N8145014) Nickel Skimmer Cone, Qty. 2 (W1026356) Nickel Sampler Cone, Qty. 2 (W1033612) Hyper Skimmer Cone, Qty. 1 (W1033995) RF Load Coil Assembly, Qty. 1 (WE021816) Support Adapter for Ball Joint Torch Injectors, Qty. 1 (W1012406) Sampler Cone Gasket, Qty. 3 (W1040148) Hyper Skimmer O-Ring, Qty. 2 (09902123) Hyper Skimmer Screw, Qty. 2 (09919737) Santoprene 1.30 mm I.D. Gray/Gray Pump Tubing, Qty. 4 (N0777444) Flared PVC 0.38 mm I.D. Green/Orange Pump Tubing, Qty. 4 (N0777042) PTFE 0.965 mm I.D. Drain Tubing, Qty. 6 (02506495) PFA 0.508 mm I.D. Sample Uptake Tubing, 12 cm Length (WE024375) Instrument Filter (left), Qty. 2 (W1036712) Instrument Filter (back right), Qty. 2 (W1036713) Spray Chamber Drain Fitting, Qty. 2 (N8145015) Nebulizer Fitting, Qty. 2 (N8145016) Male Barb CTFE Fitting, Qty. 2 (N8145017) Female Barb CTFE
Catalog No. 50-210-4760 Supplier Perkin Elmer US LLC Supplier No. N8140513
Add to Cart
Add to Cart
Request a Quote
This kit includes: Demountable Quartz Torch, Qty. 2 (N8122006) 2 mm I.D. Quartz Injector with Ball Joint, Qty. 2 (WE023948) Glass Cyclonic Spray Chamber, Qty. 1, (N8145014) Nickel Skimmer Cone, Qty. 2 (W1026356) Nickel Sampler Cone, Qty. 2 (W1033612) Hyper Skimmer Cone, Qty. 1 (W1033995) RF Load Coil Assembly, Qty. 1 (WE021816) Support Adapter for Ball Joint Torch Injectors, Qty. 1 (W1012406) Sampler Cone Gasket, Qty. 3 (W1040148) Hyper Skimmer O-Ring, Qty. 2 (09902123) Hyper Skimmer Screw, Qty. 2 (09919737) Santoprene 1.30 mm I.D. Gray/Gray Pump Tubing, Qty. 4 (N0777444) Flared PVC 0.38 mm I.D. Green/Orange Pump Tubing, Qty. 4 (N0777042) PTFE 0.965 mm I.D. Drain Tubing, Qty. 6 (02506495) PFA 0.508 mm I.D. Sample Uptake Tubing, 12 cm Length (WE024375) Instrument Filter (left), Qty. 2 (W1036712) Instrument Filter (back right), Qty. 2 (W1036713) Spray Chamber Drain Fitting, Qty. 2 (N8145015) Nebulizer Fitting, Qty. 2 (N8145016) Male Barb CTFE Fitting, Qty. 2 (N8145017) Female Barb CTFE