Learn More
Hach Company Cont Conduct Sens-Cons 0.5-SS
Catalog No. 5000207646 Shop All Hach Company Products
Encompass
0 5 cell constant-General Purpose Contacting Conductivity Sensor-3/4 inch male NPT Threaded PPS BodyNominal Cell Constant (K) 0 5 3/4 inch male NPT threaded PPS body 15m (50ft) Analog Cable Graphite Electrode
Catalog No. 50-002-07646 Supplier Hach Company Supplier No. 3433B8A50N
0 5 cell constant-General Purpose Contacting Conductivity Sensor-3/4 inch male NPT Threaded PPS BodyNominal Cell Constant (K) 0 5 3/4 inch male NPT threaded PPS body 15m (50ft) Analog Cable Graphite Electrode