Learn More
Instech Continuous flow tubing set for use with push/pull syringe pumps. .062in ID Tygon with valves. Use with HA3009 and HA3008. (610270)
SDP
Catalog No. 5081105
Encompass
The Harvard HACV062T is a continuous flow tubing set with check valves, designed for use with Harvard Apparatus models HA3009 and HA3008 (PHD Ultra series pumps configured for push/pull operation)push/pull syringe pumps. It allows for continuous, smooth flow during infusion applications.
Features
    Description: Continuous flow tubing set for use with push/pull syringe pumps.Material: The tubing is made from Tygon material.Inner Diameter: The tubing has a large inner diameter of 0.062 inches (0.062in ID), facilitating the delivery of more viscous compounds if needed.Compatibility: It is designed to work with specific Harvard Apparatus pumps, such as models HA3009 and HA3008 (PHD Ultra series pumps configured for push/pull operation).Function: When combined with a push/pull syringe pump, the valving system in the tubing set converts the reciprocating motion into a continuous flow.
Catalog No. 50-811-05 Supplier Instech Supplier No. HACV062T
The Harvard HACV062T is a continuous flow tubing set with check valves, designed for use with Harvard Apparatus models HA3009 and HA3008 (PHD Ultra series pumps configured for push/pull operation)push/pull syringe pumps. It allows for continuous, smooth flow during infusion applications.
Features
    Description: Continuous flow tubing set for use with push/pull syringe pumps.Material: The tubing is made from Tygon material.Inner Diameter: The tubing has a large inner diameter of 0.062 inches (0.062in ID), facilitating the delivery of more viscous compounds if needed.Compatibility: It is designed to work with specific Harvard Apparatus pumps, such as models HA3009 and HA3008 (PHD Ultra series pumps configured for push/pull operation).Function: When combined with a push/pull syringe pump, the valving system in the tubing set converts the reciprocating motion into a continuous flow.