Learn More
Chemplex Industries CONT ROLL POLYPROP 0.24MIL
Catalog No. 50249330
Encompass_Preferred
CHEMPLEX THIN-FILM, CONTINUOUS ROLL, Polypropylene, 6.0u (0.24mil)
Catalog No. 50-249-330 Supplier Chemplex Industries Supplier No. 425
CHEMPLEX THIN-FILM, CONTINUOUS ROLL, Polypropylene, 6.0u (0.24mil)