Learn More
Grainger Contact Block Clear 1NC 30 mm HT800
Catalog No. 502752938
Encompass_Preferred
The Eaton 14A138 is a contact block that provides a convenient method of connecting wires to Eaton contactors. It has tin-plated copper construction for corrosion resistance and has 14 AWG wire capacity. The block has a current rating of 30A at 600VAC. It has a standard 0.28" quick-connect tab spacing and is compatible with Eaton contactors like the S811 series. The block measures 1.06" x 1.06" x 0.75" and weighs approximately 0.05 lbs. It can be mounted to the contactor with a single screw, providing a secure wire termination point. The Eaton 14A138 contact block is UL/CSA recognized and ideal for a wide range of industrial control applications.
Catalog No. 50-275-2938 Supplier Grainger Supplier No. 14A138
The Eaton 14A138 is a contact block that provides a convenient method of connecting wires to Eaton contactors. It has tin-plated copper construction for corrosion resistance and has 14 AWG wire capacity. The block has a current rating of 30A at 600VAC. It has a standard 0.28" quick-connect tab spacing and is compatible with Eaton contactors like the S811 series. The block measures 1.06" x 1.06" x 0.75" and weighs approximately 0.05 lbs. It can be mounted to the contactor with a single screw, providing a secure wire termination point. The Eaton 14A138 contact block is UL/CSA recognized and ideal for a wide range of industrial control applications.