Learn More
Grainger Contact Block Clear 1NC 30 mm HT800
Catalog No. 502748869
Encompass_Preferred
The Eaton Contact Block is a 30mm panel mount contact block that provides a reliable connection point for control wiring. This Eaton contact block has screw clamp terminals that accept wires from 0.5mm2 to 2.5mm2. It is rated for voltages up to 1000V AC/1500V DC and currents up to 14A. The contact block has a thermoplastic housing that is gray in color. It can be panel mounted using the included hardware. This Eaton contact block is ideal for connecting wiring in industrial control panels and machinery.
Catalog No. 50-274-8869 Supplier Grainger Supplier No. 14A136
The Eaton Contact Block is a 30mm panel mount contact block that provides a reliable connection point for control wiring. This Eaton contact block has screw clamp terminals that accept wires from 0.5mm2 to 2.5mm2. It is rated for voltages up to 1000V AC/1500V DC and currents up to 14A. The contact block has a thermoplastic housing that is gray in color. It can be panel mounted using the included hardware. This Eaton contact block is ideal for connecting wiring in industrial control panels and machinery.