Learn More
Grainger Contact Block Clear 1NO 30 mm HT800
Catalog No. 502752925
Encompass_Preferred
The Eaton Contact Block is a 30mm panel mount contact block that provides a reliable connection point for control wiring. It features nickel-plated brass stab construction for corrosion resistance and has a current rating of 15A at 300VAC. This contact block has 12 positions and accepts wire sizes from 22-12 AWG. It can be mounted on panels up to 6mm thick using the included hardware. The Eaton contact block is UL/CSA recognized and ideal for industrial control panels and machinery requiring a robust and code-compliant power distribution block.
Catalog No. 50-275-2925 Supplier Grainger Supplier No. 14A135
The Eaton Contact Block is a 30mm panel mount contact block that provides a reliable connection point for control wiring. It features nickel-plated brass stab construction for corrosion resistance and has a current rating of 15A at 300VAC. This contact block has 12 positions and accepts wire sizes from 22-12 AWG. It can be mounted on panels up to 6mm thick using the included hardware. The Eaton contact block is UL/CSA recognized and ideal for industrial control panels and machinery requiring a robust and code-compliant power distribution block.