Learn More
Hach Company Contact Condct Alg Sens-Con0.5
Catalog No. 5000207994 Shop All Hach Company Products
Encompass
Contacting-Conductivity Analog Sensor-Cell Constant 0 5-3/4in NPT 316 Stainless Steel Compression FittingNominal Cell Constant (K) 0 5 3/4 inch NPT 316 Stainless Steel Compression Fitting 316 Stainless Steel Junction Box with 3/4in NPT Conduit Connection - Requires 1W1100 Interconnect Cable Titanium Electrode
Catalog No. 50-002-07994 Supplier Hach Company Supplier No. 3422B4D
Contacting-Conductivity Analog Sensor-Cell Constant 0 5-3/4in NPT 316 Stainless Steel Compression FittingNominal Cell Constant (K) 0 5 3/4 inch NPT 316 Stainless Steel Compression Fitting 316 Stainless Steel Junction Box with 3/4in NPT Conduit Connection - Requires 1W1100 Interconnect Cable Titanium Electrode