Learn More
Hach Company Contact Condct Alg Sens-Con0.5
Catalog No. 5000208005 Shop All Hach Company Products
Encompass
Contacting Conductivity Analog Sensor-Cell Constant 0 5-3/4 inch NPT-Stainless Steel Compression FittingNominal Cell Constant (K) 0 5 3/4 inch NPT 316 Stainless Steel Compression Fitting 6m (20 ft) Analog Cable Titanium Electrode
Catalog No. 50-002-08005 Supplier Hach Company Supplier No. 3422B4A
Contacting Conductivity Analog Sensor-Cell Constant 0 5-3/4 inch NPT-Stainless Steel Compression FittingNominal Cell Constant (K) 0 5 3/4 inch NPT 316 Stainless Steel Compression Fitting 6m (20 ft) Analog Cable Titanium Electrode