Learn More
Hach Company Contact Conduc. Dig. Sen. 0.05
Catalog No. 5000208045 Shop All Hach Company Products
Encompass
Contacting Conductivity Digital Sensor-0 05 Cell Constant-Compression Fitting StyleNominal Cell Constant (K) 0 05 1/2 inch NPT PVDF Compression Fitting 7m (23 ft) Digital Cable Titanium Electrode
Catalog No. 50-002-08045 Supplier Hach Company Supplier No. D3422A1
Contacting Conductivity Digital Sensor-0 05 Cell Constant-Compression Fitting StyleNominal Cell Constant (K) 0 05 1/2 inch NPT PVDF Compression Fitting 7m (23 ft) Digital Cable Titanium Electrode