Learn More
Hach Company Contact Conduc. Dig. Sen. 0.5
Catalog No. 5000208046 Shop All Hach Company Products
Encompass
Contacting Conductivity Digital Sensor-0 50 Cell Constant-Compression Fitting StyleNominal Cell Constant (K) 0 5 3/4 inch NPT PVDF Compression Fitting 7m (23 ft) Digital Cable Titanium Electrode
Catalog No. 50-002-08046 Supplier Hach Company Supplier No. D3422B3
Contacting Conductivity Digital Sensor-0 50 Cell Constant-Compression Fitting StyleNominal Cell Constant (K) 0 5 3/4 inch NPT PVDF Compression Fitting 7m (23 ft) Digital Cable Titanium Electrode