Learn More
Hach Company Contact Conduc. Dig. Sen. 1
Catalog No. 5000208039 Shop All Hach Company Products
Encompass
Digital Contacting Conductivity Sensor-Compression Fitting Style-Cell Constant 1 0Nominal Cell Constant (K) 1 0 3/4 inch NPT PVDF Compression Fitting 7m (23 ft) Digital Cable Titanium Electrode
Catalog No. 50-002-08039 Supplier Hach Company Supplier No. D3422C3
Digital Contacting Conductivity Sensor-Compression Fitting Style-Cell Constant 1 0Nominal Cell Constant (K) 1 0 3/4 inch NPT PVDF Compression Fitting 7m (23 ft) Digital Cable Titanium Electrode