Learn More
Hach Company Contact Conduc. Dig. Sen. 5
Catalog No. 5000208081 Shop All Hach Company Products
Encompass
Digital Contacting Conductivity Sensor-Compression Fitting Style-Cell Constant 5 0Nominal Cell Constant (K) 5 0 3/4 inch NPT PVDF Compression Fitting 7m (23ft) Digital Cable Titanium Electrode
Catalog No. 50-002-08081 Supplier Hach Company Supplier No. D3422D3
Digital Contacting Conductivity Sensor-Compression Fitting Style-Cell Constant 5 0Nominal Cell Constant (K) 5 0 3/4 inch NPT PVDF Compression Fitting 7m (23ft) Digital Cable Titanium Electrode