Learn More
Hach Company Contact Conduc. Dig. Sen. PPS
Catalog No. 5000208042 Shop All Hach Company Products
Encompass
PPS Body General Purpose Digital Contacting Conductivity Sensor for low conductivity (k 0 5)Nominal Cell Constant (K) 0 5 3/4 inch NPT Threaded PPS Body 6m (20 ft) Analog Cable Graphite Electrode Includes Digital Gateway and 1m (3 3 ft) Digital Extension Cable
Catalog No. 50-002-08042 Supplier Hach Company Supplier No. D3433B8
PPS Body General Purpose Digital Contacting Conductivity Sensor for low conductivity (k 0 5)Nominal Cell Constant (K) 0 5 3/4 inch NPT Threaded PPS Body 6m (20 ft) Analog Cable Graphite Electrode Includes Digital Gateway and 1m (3 3 ft) Digital Extension Cable