Learn More
Hach Company Contact Conduct Alg Sens-Con 1
Catalog No. 5000207335 Shop All Hach Company Products
Encompass
Contacting-Conductivity Analog Sensor-Cell Constant 1 0-3/4 inch NPT Stainless Steel Compression FittingNominal Cell Constant (K) 1 0 3/4 inch NPT 316 Stainless Steel Compression Fitting 6m (20 ft) Analog Cable Titanium Electrode
Catalog No. 50-002-07335 Supplier Hach Company Supplier No. 3422C4A
Contacting-Conductivity Analog Sensor-Cell Constant 1 0-3/4 inch NPT Stainless Steel Compression FittingNominal Cell Constant (K) 1 0 3/4 inch NPT 316 Stainless Steel Compression Fitting 6m (20 ft) Analog Cable Titanium Electrode