Learn More
Hach Company Contact Conduct Alg Sens-Con 1
Catalog No. 5000207610 Shop All Hach Company Products
Encompass
Contacting-Conductivity Analog Sensor-Cell Constant 1 0-3/4in NPT Stainless Steel Compression FittingNominal Cell Constant (K) 1 0 3/4 inch NPT 316 Stainless Steel Compression Fitting 6m (20ft) Analog Cable Titanium Electrode Generic ID
Catalog No. 50-002-07610 Supplier Hach Company Supplier No. 3422C4A20G
Contacting-Conductivity Analog Sensor-Cell Constant 1 0-3/4in NPT Stainless Steel Compression FittingNominal Cell Constant (K) 1 0 3/4 inch NPT 316 Stainless Steel Compression Fitting 6m (20ft) Analog Cable Titanium Electrode Generic ID