Learn More
Hach Company Contact Conduct Alg Sens-Con 1
Catalog No. 5000207655 Shop All Hach Company Products
Encompass
Contacting-Conductivity Analog Sensor-Cell Constant 1 0-3/4in NPT PVDF Compression FittingNominal Cell Constant (K) 1 0 3/4 inch NPT PVDF Compression Fitting Aluminum Junction Box with 3/4in NPT Conduit Connection - Requires 1W1100 Interconnect Cable Titanium Electrode
Catalog No. 50-002-07655 Supplier Hach Company Supplier No. 3422C3C
Contacting-Conductivity Analog Sensor-Cell Constant 1 0-3/4in NPT PVDF Compression FittingNominal Cell Constant (K) 1 0 3/4 inch NPT PVDF Compression Fitting Aluminum Junction Box with 3/4in NPT Conduit Connection - Requires 1W1100 Interconnect Cable Titanium Electrode