Learn More
Hach Company Contact Conduct Alg Sens-Con 5
Catalog No. 5000207453 Shop All Hach Company Products
Encompass
Contacting-Conductivity Analog Sensor-Cell Constant 5 0-3/4 inch PVDF Compression FittingNominal Cell Constant (K) 5 0 3/4 inch NPT PVDF Compression Fitting Aluminum Junction Box with 3/4in NPT Conduit Connection - Requires 1W1100 Interconnect Cable Titanium Electrode
Catalog No. 50-002-07453 Supplier Hach Company Supplier No. 3422D3C
Contacting-Conductivity Analog Sensor-Cell Constant 5 0-3/4 inch PVDF Compression FittingNominal Cell Constant (K) 5 0 3/4 inch NPT PVDF Compression Fitting Aluminum Junction Box with 3/4in NPT Conduit Connection - Requires 1W1100 Interconnect Cable Titanium Electrode