Learn More
Hach Company Contact Conduct Alg Sens-Con 5
Catalog No. 5000207656 Shop All Hach Company Products
Encompass
Contacting Conductivity Sensor-5 0 cell constant-3/4 inch NPT PVDF Compression FittingNominal Cell Constant (K) 5 0 3/4 inch NPT PVDF Compression Fitting 15m (50ft) Analog Cable Titanium Electrode
Catalog No. 50-002-07656 Supplier Hach Company Supplier No. 3422D3A50N
Contacting Conductivity Sensor-5 0 cell constant-3/4 inch NPT PVDF Compression FittingNominal Cell Constant (K) 5 0 3/4 inch NPT PVDF Compression Fitting 15m (50ft) Analog Cable Titanium Electrode