Learn More
Hach Company Contact Conduct Alg Sens-Con10
Catalog No. 5000207651 Shop All Hach Company Products
Encompass
Contacting-Conductivity Analog Sensor-Cell Constant 10 0Nominal Cell Constant (K) 10 0 Extended Sensor Body with 316 Stainless Steel Outer Electrode (requires 316 SS Ball Valve Assembly MH114M3C) 6m (20 ft) Analog Cable Titanium Electrode
Catalog No. 50-002-07651 Supplier Hach Company Supplier No. 3422E6A
Contacting-Conductivity Analog Sensor-Cell Constant 10 0Nominal Cell Constant (K) 10 0 Extended Sensor Body with 316 Stainless Steel Outer Electrode (requires 316 SS Ball Valve Assembly MH114M3C) 6m (20 ft) Analog Cable Titanium Electrode