Learn More
Hach Company Contact Conduct Alg Sens-Con10
Catalog No. 5000207736 Shop All Hach Company Products
Encompass
Contacting-Conductivity Analog Sensor-Cell Constant 10 0-Titanium ElectrodeNominal Cell Constant (K) 10 0 3/4 inch NPT 316 Stainless Steel Compression Fitting 6m (20ft) Analog Cable Titanium Electrode Engraved Number on Sensor Body
Catalog No. 50-002-07736 Supplier Hach Company Supplier No. 3422E4A20E
Contacting-Conductivity Analog Sensor-Cell Constant 10 0-Titanium ElectrodeNominal Cell Constant (K) 10 0 3/4 inch NPT 316 Stainless Steel Compression Fitting 6m (20ft) Analog Cable Titanium Electrode Engraved Number on Sensor Body