Learn More
Hach Company Contact Conduct Alg Sens-Low
Catalog No. 5000207451 Shop All Hach Company Products
Encompass
Contacting Conductivity Sensor for Low Conductivity-K 0 05 with 1/2in Stainless Steel Compression FittingNominal Cell Constant (K) 0 05 1/2 inch NPT 316 Stainless Steel Compression Fitting 6m (20ft) Analog Cable Titanium Electrode
Catalog No. 50-002-07451 Supplier Hach Company Supplier No. 3422A2A
Contacting Conductivity Sensor for Low Conductivity-K 0 05 with 1/2in Stainless Steel Compression FittingNominal Cell Constant (K) 0 05 1/2 inch NPT 316 Stainless Steel Compression Fitting 6m (20ft) Analog Cable Titanium Electrode