Learn More
Hach Company Contact. Conduct. Dig. Gateway
Supplier:  Hach Company 6120700
Encompass
Contacting Conductivity Digital GatewayDigital Gateway to convert any of the contacting conductivity analog sensors to digital output For connecting to sc digital controllers it s used directly at the end of the analog sensor cable without extensions
Catalog No. 50-002-07148